missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureLock Retrofit Kit for XCell II™ Mini-Cell

Product Code. 10368652
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10368652 1 unit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10368652 Supplier Invitrogen™ Supplier No. EI0020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

SureLock™ mini-cell lid, gel tension wedge, and molded mini-gel buffer dam.

Converts an XCell II™ Mini-Cell into an XCell SureLock™ Mini-Cell

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment) XCell SureLock™ Mini
Gel Size Mini
Product Line XCell II, XCell SureLock
Type Xcell Retrofit Kit
Quantity 1 unit
Shipping Condition Room Temperature
Max. Voltage 1500 V

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.