Learn More
Invitrogen™ Stable DAB
Description
Designed to clearly localize tissue, DNA, mRNA, and antigens in in-situ hybridization and immunoperoxidase procedures. Stable DAB reagent contains the correct amounts of diaminobenzidine and hydrogen peroxide. Stable DAB is colored red for easy identification and does not fade, creating a permanent record of results under a standard coverslip.
Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Immunocytochemistry (ICC), Immunofluorescence (IF), In Situ Hybridization (ISH)
Order Info
Shipping condition: Wet ice
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Colorimetric |
| Product Type | Stable DAB |
| Substrate | DAB |
| Substrate Properties | Chemical Substrate |
| Substrate Type | HRP (Horseradish Peroxidase) Substrate |
| Sufficient For | 10 Assays |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.