missing translation for 'onlineSavingsMsg'
Learn More

Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Mouse)

Product Code. 10013103
Change view
Click to view available options
Quantity:
100 mL (2 Bottles)
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10013103 100 mL (2 Bottles) 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10013103 Supplier Invitrogen Supplier No. WP20006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in a proprietary stabilizing reagent.

A goat anti-mouse polyclonal alk-phos conjugate, this Secondary antibody is compatable with both chemiluminescent and chromogenic methods of immunodetection, and will identify antigens bound with a primary mouse antibody.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Quantity 100 mL (2 Bottles)
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store in refrigerator (2–8°C).
Form Liquid
Isotype IgG
Reactivity Mouse
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.