missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse SAA ELISA Kit

Product Code. 10669710
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10669710

Brand: Invitrogen™ KMA0021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ELISA

The Mouse Serum Amyloid A (Ms SAA) ELISA quantitates Ms SAA in mouse serum and plasma. The assay will exclusively recognize both natural and recombinant Ms SAA. Principle of the method The Mouse SAA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05366
Assay Range 0.062 to 0.5 μg/mL
Assay Sensitivity 0.03 μ g/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 111345, 20208
Gene Symbol Saa-1,Saa2
Incubation Time 1.25 hr.
Interassay CV 5.6%
Intraassay CV 4.4
Kit Contents Pre-coated 96 well plate, Standard/Control, Standard Dilution Buffer, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution
Label or Dye HRP
Marker Gene SAA
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 1 μL, Serum 1 μL
Storage Requirements 2°C to 8°C
Target SAA
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 15 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.