missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Rapid Tris-Glycine SDS Running Buffer (Powder)

Product Code. 31904847
Change view
Click to view available options
Quantity:
5 Packets
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31904847 5 Packets Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31904847 Supplier Invitrogen™ Supplier No. LC2678
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (Powder) delivers more than 50% faster SDS-PAGE run times compared to traditional Tris-Glycine buffers, without compromising separation performance. Compatible with all Tris-Glycine gels, it offers room temperature storage and a 2-year shelf life.

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (Powder) is designed to significantly reduce SDS-PAGE run times while maintaining separation performance. By enabling separations over 50% faster than traditional Tris-Glycine running buffers, this rapid buffer supports more efficient protein electrophoresis without compromising band resolution. Compatible with all Tris-Glycine gels, including both precast and handmade formats, the powder form offers flexible preparation, room temperature storage, a two-year shelf life, and is designed for convenient routine use.

Features and benefits include:

  • More than 50% faster SDS-PAGE separations—significantly reduces electrophoresis run times compared to traditional Tris-Glycine running buffers, accelerating protein analysis workflows
  • Maintains separation performance—faster run times achieved with minimal impact to band resolution and overall separation quality
  • Compatible with all Tris-Glycine gels—designed for use with Tris-Glycine gel systems, including both Novex Tris-Glycine and SureCast handcast gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify laboratory workflows
  • Room temperature storage and long shelf life—powder format enables convenient storage with a two-year shelf life for improved inventory management
TRUSTED_SUSTAINABILITY

Specifications

Color White
Format Powder
Gel Type Tris-Glycine
Formulation Rapid Tris-Glycine SDS Running Buffer
For Use With (Application) SDS-PAGE
Quantity 5 Packets
Storage Requirements Room Temperature
Sufficient For Preparing 5 L Running Buffer
Product Line Novex
Product Type SDS Running Buffer
For Use With (Equipment) Electrophoresis Chambers
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.