missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Rapid Tris-Glycine SDS Running Buffer (10X, 500mL)

Product Code. 31904519
Change view
Click to view available options
Quantity:
5 Packets
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31904519 5 Packets 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31904519 Supplier Invitrogen™ Supplier No. LC2679
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (10X, 500 mL) delivers over 50% faster SDS-PAGE run times than traditional Tris-Glycine buffers without compromising separation performance. Supplied as a ready-to-use liquid with a 12-month shelf life, stored at 2°C to 8°C.

Invitrogen Novex Rapid Tris-Glycine SDS Running Buffer (10X, 500 mL) is designed to significantly reduce SDS-PAGE run times while maintaining separation performance. By enabling separations over 50% faster than traditional Tris-Glycine running buffers, this rapid buffer supports more efficient protein electrophoresis without compromising band resolution. Supplied as a ready-to-use liquid concentrate, the 10X format allows convenient dilution and consistent preparation, with a 12-month shelf life when stored refrigerated at 2–8°C.

Features and benefits include:

  • More than 50% faster SDS-PAGE separations—significantly reduces electrophoresis run times compared to traditional Tris-Glycine running buffers, accelerating protein analysis workflows
  • Maintains separation performance—faster run times achieved with minimal impact to band resolution and overall separation quality
  • Compatible with all Tris-Glycine gels—designed for use with Tris-Glycine gel systems, including both Novex Tris-Glycine and SureCast handcast gels
  • Concentrated 10X format—ready-to-use liquid allows simple dilution and consistent buffer preparation
  • Refrigerated storage with defined shelf life—liquid format requires storage at 2–8°C and offers a 12-month shelf life for reliable routine use
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Format Bottle
Gel Type Tris-Glycine
Volume (Metric) 500 mL
Formulation Rapid Tris-Glycine SDS Running Buffer
For Use With (Application) SDS-PAGE
Quantity 5 Packets
Storage Requirements Store at 2°C to 8°C.
Sufficient For Preparing 5 L Running Buffer
Product Line Novex
Product Type SDS Running Buffer
For Use With (Equipment) Electrophoresis Chambers
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.