missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex RANTES Human ELISA Kit

Product Code. 10018802
Change view
Click to view available options
Quantity:
96 Assays
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10018802 96 Assays 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10018802 Supplier Invitrogen™ Supplier No. KAC1511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Designed to quantify protein levels of human RANTES in plasma, supernatant, and other biological fluids

Regulated upon Activation Normal T cell Expressed & Secreted (RANTES) is part of the chemokine superfamily.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Plasma, Supernatant
For Use With (Application) Colorimetric
Gene ID (Entrez) 6352
Kit Contents Antibody-HRP, Chromogen, Controls, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Wash buffer
Precision 2pg/mL
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Target Chemokine (C-C motif) ligand 5
Test Time 2.5 hr.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.