Learn More
Invitrogen™ Human Prostaglandin E2 ELISA Kit
Description
The Human Prostaglandin E2 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (ELISA). This assay is designed to detect and quantify the level of human prostaglandin E2 in biological samples. Principle of the method This assay is based on the competition between Prostaglandin E 2 (PGE2) and a PGE2 alkaline phosphatase tracer for a limited amount of PGE2 specific monoclonal antibody. Because of the competition between PGE2 in the sample and PGE2 tracer for the PGE2 antibody, the signal obtained with the assay will be inversely proportional to the amount of PGE 2 in each sample. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | NM |
| Assay Range | 31.3 to 4000 pg/mL |
| Conjugate | AP |
| Product Type | ELISA |
| Sample Type | Plasma, Supernatant, Urine |
| Target Species | Human |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Incubation Time | 3 hr. |
| Interassay CV | 11.7% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.