missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT5 alpha (Total) Human ELISA Kit

Product Code. 10637035
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10637035

Brand: Invitrogen™ KHO0751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

STAT5a Antibody-Coated 96-Well Plate; STAT5a Total Standard; Standard Diluent Buffer; STAT5a Total Detection Antibody; anti-rabbit IgG-HRP (100X); HRP Diluent; Wash Buffer Concentrate (25X); Stabilized Chromogen, TMB; Stop Solution; Plate Covers; Detailed protocol with validation tests

Sandwich ELISA Kit

Kit is designed to detect and quantify the level of STAT5 alpha (Total) in fresh or frozen human cell lysates. Cross-reactivity has been observed in mouse and rat cells. The assay recognizes both natural and recomb. human STAT5 alpha (Total), independent of its phosphorylation state. Principle of the method A monoclonal capture antibody specific for human STAT5 alpha (Total) has been coated onto wells of 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a HRP labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the O.D. can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42229,P42230,Q62771
Assay Range 1.56-100 ng/mL
Assay Sensitivity <1 ng/mL
Concentration 1.56 to 100ng/mL
Conjugate HRP
Product Type ELISA Kit
Sample Type Cell Lysates
Substrate Type Transcription Factors
Target Species Human, Mouse, Rat
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20850, 24918, 6776
Gene Symbol MGF,STAT5
Incubation Time 4 hr.
Interassay CV 7.3%
Intraassay CV 4.5%
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene STAT5a
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate,10 μL
Storage Requirements 2°C to 8°C
Target Human, Mouse, Rat
Test Time 1 hr. 20 min.
Total Assay Time 4 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.