Learn More
Invitrogen™ Novex™ phosphoELISA™ GSK-3β [pS9] ELISA Kit
Description
Includes
β Coated 96-Well Plate, GSK-3β [pS9] Standard, Standard Diluent Buffer, GSK-3β [pS9] Detection Antibody, Anti-Rabbit IgG-HRP (100X), HRP Diluent, Wash Buffer Concentrate (25X), Stabilized Chromogen, TMB, Stop Solution, Plate Covers, Detailed protocol with validation tests
The GSK-3β [pS9] ELISA research use only kit is intended for the detection of GSK-3β [pS9] from human cell lysates using 96-well plates and a microplate reader.
- Easy to run Sandwich ELISA
- More sensitive than westerns
- Convenient precoated, removable 8-well strips
- Detect Low-Expressing Proteins and Use Less Sample
- Convenient Kit Format
- Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency
Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Order Info
Shipping condition: Wet ice
Specifications
Specifications
| Assay Sensitivity | 1.0U/mL |
| Concentration | 1.6U/mL |
| Conjugate | HRP |
| Sample Type | Cell Extracts |
| Substrate Type | Serine/Threonine Kinases |
| Target Species | Mouse, Rat |
| Gene ID (Entrez) | 2932 |
| Incubation Time | 4 hr. |
| Marker Gene | GSK-3β |
| Quantity | 96 Assays |
| Show More |
For research use only. Not intended for human or animal therapeutic or diagnostic use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.