missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CREB (Phospho) [pS133] ELISA Kit

Product Code. 10161253
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10161253 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10161253 Supplier Invitrogen™ Supplier No. KHO0241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ELISA

Kit is designed to detect and quantify the level of CREB (Phospho) [pS133] in fresh or frozen human cell lysates. Cross-reactivity has been observed in mouse cells. The assay recognizes both natural and recombinant CREB (Phospho) [pS133]. Principle of the method A monoclonal capture antibody specific for human CREB (Phospho) [pS133] has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CREB (Cyclic AMP response element binding protein) is a 43 kDa basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation of CREB occurs via p44/42 MAP kinase and p90RSK, and via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB is involved in different cellular processes including the synchronization of circadian rhythmicity, differentiation of adipose cells, and learning and memory. In humans, the gene is located on the q arm of chromosome 2. Diseases associated with CREB dysfunction include Alzheimer’s Disease, histiocytoma and soft tissue melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16220,Q01147
Assay Range 0.8 to 50 U/mL
Assay Sensitivity <0.9 U/mL
Concentration 1.6 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Substrate Type Transcription Factors
Target Species Mouse, Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 12912, 1385
Gene Symbol CREB,CREB-1
Incubation Time 4 hr.
Interassay CV 5.8%
Intraassay CV 4
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene CREB
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Storage Requirements 2°C to 8°C
Target CREB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.