missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ Tris-Acetate Mini Protein Gels, 3 to 8%, 1.0 mm, 2D-well
Description
Includes
One box of 10 gels
- Can also be run with Tris-Glycine Native Running Buffer to resolve native proteins more effectively than with the Tris-Glycine gel system
- Made with high-purity, strictly quality-controlled reagents: Tris base, acetic acid, acrylamide, bis-acrylamide, TEMED, APS, and highly purified water
- Do not contain SDS
2D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 6 months. |
| Gel Percentage | 3 to 8% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Tris-Acetate |
| Separation of | Protein |
| Separation Range | 36 to 500 kDa |
| Separation Type | Denaturing, Native |
| Wells | 2D-well |
| Mode of Separation | Molecular Weight |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?