missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Recombinant Protein G Agarose

Product Code. 11598896
Change view
Click to view available options
:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11598896 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11598896 Supplier Invitrogen™ Supplier No. 15920010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

rProtein G Agarose is supplied as a 50% suspension in PBS (pH 7.0), 20% ethanol.

For the purification of IgG from most mammalian species and does not bind IgM, IgA, IgE or serum albumin

  • Recombinant Protein G (rProtein G) Agarose is a ready-to-use affinity chromatography matrix supplied as a 50% slurry (in phosphate buffered saline (PBS), pH 7.0; 20% ethanol)
  • The rProtein G is a recombinant form of streptococcal protein G engineered to eliminate nonspecific binding to serum albumin, and is immobilized on 4% agarose beads
  • rProtein G Agarose has a binding capacity of approximately 20mg of human IgG permL of beads
  • rProtein G Agarose binds to a broader range of IgG isotypes than react with protein A
  • The binding characteristics and stability of rProtein G Agarose facilitate rapid and repeated use for affinity chromatography
  • To purify IgG, load crude samples (serum, ascites fluid, or hybridoma supernatant) onto a column of rProtein G Agarose under physiologic conditions of salt and pH
  • Non-IgG components do not bind to the beads and elute in the flow through fraction; Purified IgG is then eluted with a low pH buffer

PCR, PCR and Real-Time PCR

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Column Type Affinity Column
Content And Storage rProtein G Agarose is supplied as a 50% suspension in PBS (pH 7.0), 20% ethanol. Store at +4°C.
Separation of Protein
Form Suspension
Shipping Condition Wet Ice
Stationary Phase Protein G
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.