missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein A Agarose
Description
Includes
Protein A Agarose is supplied as a 45% to 55% suspension in 20mM sodium phosphate (pH 7.2), 0.15M NaCl, 0.02% (w/v) merthiolate.
- The matrix consists of purified Protein A covalently attached to cross-linked 4% agarose beads using the cyanogen bromide method
- Protein A Agarose can bind >18mg of human IgG/mL of gel
- Binding capacity for human IgG is determined by affinity chromatography
- Protein A interacts with some classes of IgM and IgA
PCR, PCR and Real-Time PCR
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Column Type | Affinity Column |
| Content And Storage | Protein A Agarose is supplied as a 45% to 55% suspension in 20 mM sodium phosphate (pH 7.2), 0.15 M NaCl, 0.02% (w/v) merthiolate. Store at +4°C. |
| Form | Suspension |
| Shipping Condition | Wet Ice |
| Stationary Phase | Protein A |
| Product Type | Protein A Agarose |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Individuate un'opportunità di miglioramento?