missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIP-2 (CXCL2) Rat ELISA Kit

Product Code. 10242403
Change view
Click to view available options
Quantity:
96 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10242403 96 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10242403 Supplier Invitrogen™ Supplier No. KRC1021

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sandwich ELISA Kit

The Rat MIP-2 (CXCL2) ELISA quantitates Rt MIP-2 in human serum, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt MIP-2. Principle of the method The Rat MIP-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Macrophage inflammatory protein 2 is a chemokine involved in the pulmonary inflammatory response. MIP-2 is produced by activated monocytes and neutrophils and expressed at sites of inflammation. In vitro, it suppresses hematopoietic progenitor cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P30348
Assay Range 10-640 pg/mL
Assay Sensitivity <1 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Serum, Supernatant
Target Species Rat
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 114105
Gene Symbol Cxcl2
Incubation Time 3.5 hr.
Interassay CV 8.6%
Intraassay CV 5.9%
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene MIP-2
Quantity 96 Assays
Regulatory Status RUO
Sample Volume Serum (50 μL), Supernatants (50 μL)
Storage Requirements 2°C to 8°C
Target Rat
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.