missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ MCP-1 Mouse ELISA Kit

Product Code. 10170573
Change view
Click to view available options
Quantity:
192 Assays
96 Assays
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10170573 96 Assays Each
10272393 192 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10170573 Supplier Invitrogen™ Supplier No. KMC1011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Quantifies natural and recombinant mouse Monocyte Chemoattractant Protein-1 (MCP-1) in serum, buffered solution, or cell culture medium

MCP-1 attracts recruits monocytes, memory T cells, and dendritic cells to sites of tissue injury and infection.

Applications:

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_011333.3, NP_035463.1
Assay Sensitivity <9pg/mL
Sample Type Serum, Supernatant
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) 20296
Incubation Time 3.8 hr.
Kit Contents Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer
Marker Gene MCP-1
Quantity 96 Assays
Target chemokine (C-C motif) ligand 2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.