missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ MCP-1 Human ELISA Kit

Product Code. 10018992
Change view
Click to view available options
Quantity:
96 Assays
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10018992 96 Assays 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10018992 Supplier Invitrogen™ Supplier No. KHC1011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accession Number NM_002982.3, NP_002973.1
Assay Sensitivity <20pg/mL
Sample Type Plasma, Serum, Supernatant
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) 6347
Incubation Time 3 hr.
Kit Contents Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer
Marker Gene MCP-1
Quantity 96 Assays
Target chemokine (C-C motif) ligand 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.