missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ MCP-1 Human ELISA Kit
Click to view available options
Quantity:
192 Assays
Unit Size:
192 tests
Description
MCP-1 attracts recruits monocytes, memory T cells, and dendritic cells to sites of tissue injury and infection.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Specifications
| Accession Number | NM_002982.3, NP_002973.1 |
| Assay Sensitivity | <20pg/mL |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Microplate Reader |
| Gene ID (Entrez) | 6347 |
| Incubation Time | 3 hr. |
| Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Marker Gene | MCP-1 |
| Quantity | 192 Assays |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction