missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminex™ IL-10 Human Singleplex Bead Kit

Product Code. 10100383
Click to view available options
Quantity:
100 Assays
Unit Size:
100 tests
This item is not returnable. View return policy

Product Code. 10100383

Brand: Invitrogen™ LHC0101

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.

Requires

Buffer reagents needed to complete the reaction are sold separately under Catalog number LHB0001.

For the measurement of human IL-10 (Interleukin-10) in serum, plasma or tissue culture supernatant

The assay may be run alone or in combination with other Human Singleplex Bead Kits. These reagents are intended for use in the Luminex™ 100™ or 200™ System only.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Assays
Assay Sensitivity <5 pg/mL
For Use With (Application) Fluorescent Detection
Gene ID (Entrez) 3586
Protein Subtype Interleukin 10
Sample Type Cell Culture Supernatant, Plasma, Serum
Kit Contents Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard
Regulatory Status For research use only (RUO)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.