missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminex™ Cytokine Mouse Magnetic 20-Plex Panel

Product Code. 15752516
Change view
Click to view available options
Quantity:
100 Assays
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15752516 100 Assays Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752516 Supplier Invitrogen™ Supplier No. LMC0006M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains premixed antibody coated capture beads, standard, premixed detector antibody, diluents, SAV concentrate, buffers, wash solution, filter plate, flat bottom plate, complete protocol and lot-specific technical data sheet.

Specifically designed for quantifying mouse cytokines, chemokines, and growth factors in serum, plasma, and tissue culture supernatant

By measuring 20 analytes simultaneously, the assay panel provides more data from each sample, saving costs and time. This assay may be run alone or in combination with other select Luminex singleplex kits. This assay may be run alone or in combination with other select Luminex singleplex kits. The panel employs magnetic beads, facilitating automation, decreasing hands-on time, and increasing throughput and precision. Suitable for use with Luminex™ 100/200, FLEXMAP 3D™ and MAGPIX™ systems.

Largest comprehensive mouse analyte panel
The Mouse Cytokine Magnetic 20-Plex Panel for the Luminex platform provides reagents for the accurate, reproducible, and sensitive quantitation of mouse proteins including: GM-CSF, IP-10, FGF-basic, IFN-γ, VEGF, IL-1α, MCP-1, IL-1β, MIG, IL-2, MIP-1α, IL-4, IL-5, IL-6, IL-10, IL-12 (p40/p70), IL-13, IL-17, and TNF-α. Luminex xMAP technology—effective and proven analytical tool
The Mouse Cytokine Magnetic 20-Plex Panel is based on xMAP; technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. At the completion of the sandwich immunoassay, beads can be read using the Luminex 200™ or FLEXMAP 3D detection systems, instruments based on flow cytometry, two lasers, real-time digital signal processing, and xPONENT™ software to distinguish bead color (analyte) and assay signal strength (PE) fluorescence intensity.

MAGPIX system—brings the power of multiplex to every lab
The MAGPIX system may also be used for sample analysis. This instrument utilizes xMAP technology and is built on light-emitting diodes (LEDs) for excitation, a CCD camera for bead and analyte detection, and high-speed digital signal processors. The MAGPIX system is more compact and robust, providing a very cost effective magnetic bead-based multiplexing tool. Simple, out-of-the-box set-up allows researchers to rapidly perform singleplex or multiplex assays of up to 50 targets in single biological samples. The MAGPIX system is easy to use—ideal for both new and experienced users alike.

Magnetic beads let you do more with your time
Magnetic bead–based assays use Luminex MagPlex magnetic microspheres. All other assay components are the same as the equivalent Luminex polystyrene bead–based assays providing identical quality and consistency performance. MagPlex technology takes advantage of magnetic properties to simplify assay wash steps and maximize uniformity of results. Magnetic bead-based assays also enable automation, decrease hands-on time, and increase throughput and precision. Easy-to-use protocols allow you to get your results in as little as 3.5 hours. In addition, these assays are compatible with both vacuum and magnetic wash stations and work well with the MAGPIX system as well as other Luminex xMAP platforms. More validation for confidence in your results
The Mouse Cytokine Magnetic 20-Plex Panel has been validated using the same rigorous criteria that distinguishes Luminex assays. Each kit supplied with a product insert that outlines the specifications for each marker in the assay.

  • Superior performance: accurate, reproducible, and sensitive quantitation of multiple proteins
  • High quality: fully qualified antibodies permit excellent specificity and sensitivity
  • Fast and easy protocols: perform your multiplex assay and analyze your data typically in less than one day
    • Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Assays
Bead Type Magnetic
For Use With (Application) Fluorescent Detection
For Use With (Equipment) Luminex Instruments
Gene ID (Entrez) 14182, 12981, 15978, 16175, 16176, 16183, 16189, 16191, 16193, 16153, 16163, 16160, 16171, 15945, 14825, 20296, 17329, 20302, 21926, 22339
Protein Subtype FGF-Basic, GM-CSF, IFN-γ, IL-1α, IL-1β, IL-10, IL-12 (p40/p70), IL-13, IL-17, IL-2, IL-4, IL-5, IL-6, IP-10, KC, MCP-1, MIG, MIP-1α, TNF-α, VEGF
Sample Type Serum
Kit Contents Buffers, Complete protocol, Diluents, Filter plate, Flat bottom plate, Lot specific technical data sheet, Premixed antibody coated capture beads, Premixed detector antibody, SAV concentrate, Standard, Wash solution

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.