missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Automated Maintenance Plate (Luminex™ System)

Product Code. 10376269
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10376269

Brand: Invitrogen™ CN020601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Compatible with Luminex 100 and 200 Systems

Automated Maintenance Plate (AMP) allows for easy walk-away maintenance and calibration.

Cell Analysis, Immunoassays

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at RT
Product Line Luminex
Product Type Maintenance Plate
For Use With (Equipment) Luminex Instruments
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.