Learn More
Invitrogen™ Human IgG Subclass ELISA Kit
Description
The Human IgG Subclass Profile (Hu IgG) ELISA quantitates Hu IgG subclasses in human serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu igG1, IgG2, IgG3 or IgG4.
Principle of the method
The Human IL-10 US solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Serum |
| Target Species | Human |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Kit Contents | Pre-coated 96 well plates, Standard, Sample Diluent, Assay Buffer concentrate, Serum Control, MAbs to IgG Subclasses(4), Peroxidase-conjugated Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Label or Dye | HRP |
| Marker Gene | IgG1, IgG2, IgG3, IgG4 |
| Quantity | 2 x 96 Tests |
| Show More |
Safety and Handling
- Human IgG Subclass ELISA Kit
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 2-methyl-3(2H)-isothiazolone
Your input is important to us. Please complete this form to provide feedback related to the content on this product.