Learn More
Invitrogen™ IL-8 Human ELISA Kit, Ultrasensitive
A cytokine chemoattractant protein for neutrophils
£506.00 - £710.00
Specifications
Accession Number | P10145 |
---|---|
Assay Range | 0.39-25 pg/mL |
Assay Sensitivity | <100 fg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10292633
|
Invitrogen™
KHC0084 |
96 Tests |
£506.00
96 tests |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10660924
|
Invitrogen™
KHC0083 |
2 x 96 Tests |
£710.00
192 tests |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
Description
The Human Interleukin 8 (IL-8) ELISA quantitates Hu IL-8 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-8. Principle of the method The Human IL-8solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The IL-8 Human Ultrasensitive ELISA Kit quantifies natural and recombinant human IL-8 human serum, plasma, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
P10145 | |
<100 fg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 | |
5.8% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
0.39-25 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric | |
3576 | |
7.6% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
<0.1pg/mL | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Human | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.