missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex IL-6R Human ELISA Kit

Product Code. 10436673
Click to view available options
Quantity:
96 Assays
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 10436673

Brand: Invitrogen™ KHR0061

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Designed to quantify the levels of human soluble interleukin-6 receptor (IL-6R)

Determines levels in serum, plasma, buffered solution, or cell culture medium recognizing both natural and recombinant Hu sIL-6R

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Plasma, Serum, Supernatant
For Use With (Application) Colorimetric
Gene ID (Entrez) 3570
Kit Contents Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer
Precision <8pg/mL
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Target Interleukin 6 receptor
Test Time 3 hr.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.