missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-13 ELISA Kit

Product Code. 10242303
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10242303

Brand: Invitrogen™ KMC2221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ELISA

The Mouse Interleukin 13 (Ms IL-13) ELISA quantitates Ms IL-13 in mouse serum, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-13. Some cross-reactivity is observed in human and rat samples. Principle of the method The Mouse IL-13 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20109
Assay Range 3.9 to 250 pg/mL
Assay Sensitivity <2 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Serum, Supernatant
Target Species Mouse
For Use With (Application) Colorimetric
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 16163
Gene Symbol IL-13
Interassay CV 8%
Intraassay CV 6.7
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Precision <2 pg/mL
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum 10 μL, Supernatant 10 μL
Storage Requirements 2°C to 8°C
Target IL-13
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.