missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-12 p40/p70 Human ELISA Kit

Product Code. 10049272
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10049272 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10049272 Supplier Invitrogen™ Supplier No. KHC0121

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sandwich ELISA Kit

The Human Interleukin 12 p40/p70 (Hu IL-12 p40/p70) ELISA quantitates Hu IL-12 p40/p70 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-12 p40/p70. Principle of the method The Human IL-12 p40/p70 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-12 (IL-12) is a heterodimeric 70 kD (p70) cytokine composed of two covalently linked, glycosylated chains, 40kD (p40) and 35-kD (p35). Following protein synthesis, an active heterodimer (p70) and a homoodimer of p40 are formed. IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma (IFN-γ) production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. It is now established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-γ and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29459,P29460
Assay Range 7.8-500 pg/mL
Assay Sensitivity <2 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Colorimetric
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3592, 3593
Gene Symbol CLMF,CLMF2,IL-12B,IMD28,IMD29,NKSF,NKSF2,CLMF,IL-12A,NFSK,NKSF1,P35
Interassay CV 3.6%
Intraassay CV 3.2%
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Precision <2pg/mL
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 3 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.