Learn More
Invitrogen™ Novex IL-10 Mouse ELISA Kit
Brand: Invitrogen™ KMC0101
Code : 50
Additional Details : Weight : 0.70000kg
Specifications
Serum, Supernatant | |
16153 | |
<13pg/mL | |
For research use only (RUO) | |
3 hr. |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays | |
Interleukin 10 |