missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex IL-10 Human Ultrasensitive ELISA Kit
Description
The IL-10 Human Ultrasensitive ELISA Kit quantifies natural and recombinant human IL-10 human serum, plasma, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Specifications
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric |
| Gene ID (Entrez) | 3586 |
| Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Precision | <0.2pg/mL |
| Quantity | 96 Assays |
| Regulatory Status | For research use only (RUO) |
| Target | Interleukin 10 |
| Test Time | 4 hr. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?