missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ Transfer Stack, nitrocellulose, mini
GREENER_CHOICE

Product Code. 10227972
Change view
Click to view available options
Quantity:
10 sets
3 x 10 sets
Dimensions (LxW):
Mini, 8 cm x 8 cm
Unit Size:
Pack of 10
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensions (LxW) unitSize
10227972 10 sets Mini, 8 cm x 8 cm Pack of 10
12219319 3 x 10 sets Mini, 8 cm x 8 cm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10227972 Supplier Invitrogen™ Supplier No. IB301002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One box of iBlot™ Transfer Stacks containing 10 stacks.

iBlot Transfer Stacks are used to transfer proteins

The iBlotTransfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlotTransfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

Nitrocellulose membrane

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic and electrostatic interactions
  • Pore size: 0.2μm
  • Protein binding capacity: 209μg/cm2

 

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

 

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
Quantity 10 sets
Format Transfer Stack
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage 10 sets of transfer stacks in 1 box. Store at room temperature.
Shipping Condition Room Temperature
Dimensions (LxW) Mini, 8 cm x 8 cm
Product Line iBlot
Sufficient For 10 Blots
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • iBlot Transfer Stack, nitrocellulose, mini
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)-1,3-Propanediol ethylene glycol,aluminium oxide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.