missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flat Gel Loading Tips
Description
Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 μL.
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Description | Flat Tips |
| For Use With (Application) | 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis |
| For Use With (Equipment) | Universal Fit |
| Product Line | Eppendorf |
| Type | Flat Gel Loading Tip |
| Filtered | No |
| Quantity | 200/ pkg. |
| Shipping Condition | Room Temperature |
| Sterility | Non-sterile |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?