missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ G-CSF Human ELISA Kit
Description
G-CSF supports growth of hematopoietic colonies.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Specifications
| Assay Sensitivity | <20pg/mL |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Microplate Reader |
| Gene ID (Entrez) | 1440 |
| Incubation Time | 3 hr. |
| Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Marker Gene | G-CSF |
| Quantity | 192 Assays |
| Target | Granulocyte colony stimulating factor 3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?