missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EKMax™ Enterokinase
Recombinant preparation of the catalytic subunit of bovine enterokinase
£868.00 - £2650.00
Specifications
Product Type | Enterokinase |
---|---|
Species | Cattle/Bovine |
Protein Tag | His Tag |
Product Line | EKMax™ |
Includes: EKMax Enterokinase is provided with 10X buffer.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10184792
|
Invitrogen™
E18001 |
250 Units |
£868.00
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10453242
|
Invitrogen™
E18002 |
1000 Units |
£2650.00
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Recognizes the sequence Asp-Asp-Asp-Asp-Lys and cleaves peptide bond after lysine residue
- Enzyme can be used to cleave any fusion protein that carries this peptide sequence
- Well-suited to removal of fusion tags from recombinant proteins
- Unit Definition: One unit of EKMaxis amount of enzyme required to digest 20μg of thioredoxin-CAT fusion protein to 90% completion in 16 hours at 37°C; One EKMaxunit is equivalent to ∽190 trypsinogen activation units
GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
Enterokinase | |
Cattle/Bovine | |
His Tag | |
EKMax™ |
For Research Use Only. Not for use in diagnostic procedures.