Learn More
Invitrogen™ Bovine IFN gamma ELISA Kit
Description
The Bovine Interferon gamma (Bov IFN-gamma) ELISA semi-quantitates Bov IFN-gamma in plasma or cell culture medium. The assay will exclusively recognize natural Bov IFNγ. Principle of the method The Bov IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P07353 |
| Assay Range | semi-quantitative |
| Assay Sensitivity | semi-quantitative |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Supernatant |
| Target Species | Bovine |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 281237 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.