missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human c-Met (Soluble) ELISA Kit

Product Code. 10402355
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10402355 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10402355 Supplier Invitrogen™ Supplier No. KHO2031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ELISA

The Human c-Met (Soluble) ELISA quantitates Hu c-Met in human serum, plasma and cell culture supernatant. The assay will exclusively recognize both natural and recombinant Hu c-Met. Principle of the method The Human c-Met solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08581
Assay Range 0.78 to 50 ng/mL
Assay Sensitivity <0.5 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4233
Gene Symbol AUTS9,DFNB97,HGFR,RCCP2,c-Met
Incubation Time 4 hr.
Interassay CV 8.1%
Intraassay CV 6.1
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene c-Met (soluble)
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 5 μL, Serum 5 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target c-Met
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.