missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BenchMark™ Fluorescent Protein Standard

Product Code. 10377952
Change view
Click to view available options
Quantity:
125 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10377952 125 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10377952 Supplier Invitrogen™ Supplier No. LC5928
  • ONLINE EXCLUSIVE PRICE
    £372.00
    £301.65 / 125µL
    Save £70.35
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

125μL (25-50 applications) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, and Coomassie Blue G-250

Consists of fluorescent dye-conjugated proteins, ranging in size from approx. 11-155kDa

The standard proteins can be resolved into sharp bands in SDS-PAGE and visualized using a UV transilluminator or commonly used laser scanner systems.

 

  • Seven tight, clear bands that resolve well in NuPAGE™ Bis-Tris Gels and Novex Tris-Glycine Gels
  • Visualization directly under the UV light box along with the proteins labeled with the Lumio™ Green Detection Reagent
  • Easy molecular weight estimation for fluorescence-conjugated proteins in SDS-PAGE and western blots
  • A ready-to-use format
  • Detection with Coomassie™ Brilliant Blue R-250
  • Gel Compatibility: NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels

 

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Warranty: 90 days

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 125 μL (25-50 applications) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, and Coomassie™ Blue G-250.

Store at -20°C. Avoid repeated freezing and thawing. Stable for 6 months when properly stored.
Detection Method 488 nm channel
Number of Markers 7
Ready to Load Yes
Size Range 11 to 155 kDa
Gel Compatibility Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels
Molecular Weight (g/mol) 155, 98, 63, 40, 32, 21, 11 kDa
Quantity 125 μL
Shipping Condition Dry Ice
Product Line BenchMark
Product Type Protein Ladder
Stain Type Alexa Fluor 488
System Type Western Blotting, SDS-PAGE
Show More Show Less
I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?

- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.

I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?

- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.

I used one of your pre-stained standards on a Tris-Glycine gel and noticed that the molecular weights of the proteins were different than on a NuPAGE Bis-Tris gel. What is the reason for this?

Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.

I used one of your protein standards and am seeing some extra bands in the lane. Can you offer some suggestions?

- While loading, take care to make sure that there is no cross-contamination from adjacent sample lanes.
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can result in extra bands and this is a problem especially with silver-stained gels.
- Improper storage of the standard or repeated freeze/thawing can result in protein degradation.

I used one of your protein standards and the bands look non-distinct and smeary. What should I do?

Here are some suggestions:

- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can cause smearing and this is a problem especially with silver stained gels.
- Bands will not be as well resolved in low percentage gels. Try using a higher percentage gel.
- If the bands look smeary and non-distinct after a western transfer/detection, this may be due to the antibody being too concentrated. Follow the manufacturer's recommended dilution or determine the optimal antibody concentration by dot-blotting.

A couple of bands in my protein standard are missing on the gel. Can you help me troubleshoot?

Here are some suggestions:

- Check the gel type/percentage of the gel that was used. Depending on the gel type and/or percentage, all the bands may not be seen. For example, the smallest bands of the protein standard may not resolve on a very low percentage gel whereas the higher molecular weight bands may not resolve on a high percentage gel.
- Check the expiration date on the protein standard. Expired lots may result in faded or missing bands due to protein degradation.
- Check the storage conditions for the protein standard. Improper storage conditions will compromise the stability of the proteins in the standard.
- Make sure that the protein standard was not heated/boiled prior to loading on the gel. Our protein standards are ready to load and we do not recommend heating/boiling them as this may cause degradation of proteins in the standard.

What is the identity of each protein band in the BenchMark Fluorescent Protein Standard?

The identity of each protein band in the BenchMark Fluorescent Protein Standard is proprietary.

Can I use the BenchMark Fluorescent Protein Standard for protein quantitation?

The concentration of each protein in the standard is proprietary. We do not recommend using the standard for quantitation purpose.

What are the maximum excitation and emission wavelengths for the Alexa Fluor 488 dye that is bound to the proteins in the BenchMark Fluorescent Protein Standard?

The maximum excitation of the dye is at 493 nm and the maximum emission is at 516 nm.

What are the apparent molecular weights of the proteins in the BenchMark Fluorescent Protein Standard?

The BenchMark Fluorescent Protein Standard is designed for visualization of molecular weight ranges of proteins labeled with Lumio Green Detection kit (Cat. No. LC6090) or other fluorescent-conjugated proteins. The standard consists of a set of wide molecular weight range proteins conjugated to a fluorescent dye, Alexa Fluor 488. The standard can also be detected with Coomassie staining. It is suitable for use with NuPAGE or Tris-Glycine gels. Here are the apparent molecular weights of the proteins in the BenchMark Fluorescent Protein Standard in the different gels:

- Band 1: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 155 kda; 4-20% Tris-Glycine gel - 155 kDa
- Band 2: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 98 kDa; 4-20% Tris-Glycine gel - 100 kDa
- Band 3: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 63 kDa; 4-20% Tris-Glycine gel - 65 kDa
- Band 4: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 40 kDa; 4-20% Tris-Glycine gel - 41 kDa
- Band 5: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 32 kDa; 4-20% Tris-Glycine gel - 33 kDa
- Band 6: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 21 kDa; 4-20% Tris-Glycine gel - 23 kDa
- Band 7: NuPAGE 4-12% Bis-Tris gel with MES/MOPS buffer - 11 kDa; 4-20% Tris-Glycine gel - 12 kDa

What are the storage conditions and shelf life for the BenchMark Fluorescent Protein Standard?

We recommend storing the BenchMark Fluorescent Protein Standard in the dark at -20 degrees C and in aliquots to avoid repeated freezing and thawing. It is stable for 6 months when properly stored.

Do protein standards run differently on a Zymogram gel compared to a regular Tris-Glycine gel?

Zymogram gels are essentially Tris-Glycine gels containing the substrate. Protein standards run based solely on the percentage of acrylamide and hence should run the same in both kinds of gels. It is quite possible though that if the standard is prestained, the proteins will appear a different color because of the staining (or pre-staining) of the Zymogram gels.

Can I use any of your protein standards for estimation of protein quantity (protein quantitation)?

Our protein standards are not designed for protein quantitation.

What is the recommended gel loading volume for your protein standards?

Please find this information in the respective manuals for the individual protein standards.

Do I need to boil your protein standards before loading on the gel?

Our protein standards are ready to load. We do not recommend heating them as this may cause protein degradation.

Do I have to add reducing agent to your protein standards?

Except for our NativeMark Unstained Protein Standard (designed for native electrophoresis), all of the other unstained and prestained standards we offer (Invitrogen Sharp, SeeBlue, SeeBlue Plus2, BenchMark, HiMark) have been pre-reduced (by a proprietary method). Hence, you do not need to add reducing agent.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.