missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Novex™ AP Chemiluminescent Substrate Enhancer (Nitro Block II™)
Description
The Novex™ AP Chemiluminescent Substrate Enhancer consists of a 20X solution of Nitro-Block-II™ intended for use in conjunction with Novex™ AP Chemiluminescent Substrate to increase the intensity of chemiluminescent signals when probing blots on nitrocellulose membranes. The enhancer produces a hydrophobic environment on the membrane which increases signal intensity and allows shorter exposure times for signal acquisition.
Specifications
Specifications
| Product Type | Substrate Enhancer |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Product Line | Novex |
| Quantity | 1 Bottle |
| Substrate | Alkaline Phosphatase |
| Content And Storage | Single bottle labeled: Novex™ AP Chemiluminescent Substrate Enhancer (Nitro Block II™). Store at 4°C |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?