Learn More
Invitrogen™ Novex™ AKT [pS473] Antibody Pair
Concentrated
Brand:  Invitrogen™ CHO0115
Product Code. 10435273
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Anti-Rabbit IgG-HRP concentrate and technical data sheet.
Requires : Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).
Description
Description
Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of phosphorylated Akt [pS473]. Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate.
Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate.
Build Your Own Immunoassay, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Drug Discovery and Development, Immunoassays, Kinase Antibodies and Immunoassays, Kinase Biology, Ready-To-Use Immunoassay, Target and Lead Identification and Validation
Order Info
Shipping Condition: Wet Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_001014431.1, NM_001014432.1, NM_005163.2, NP_001014431.1, NP_001014432.1, NP_005154.2
AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
AKT1