missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AcTEV™ Protease

Product Code. p-4921417
Click to view available options
Quantity:
10,000 U
1000 U
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10216572

Brand: Invitrogen™ 12575015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

AcTEV™ Protease is supplied with a vial of 20X TEV buffer (1M Tris-HCl (pH 8.0), 10mM EDTA) and a vial of 100mM DTT.

Specifically recognizes a seven amino acid sequence (Glu-Asn-Leu-Tyr-Phe-Gln-Gly, cleaving between Gln and Gly), ideal for removing affinity tags from fusion proteins

An improved version of Tobacco Etch Virus (TEV) AcTEV™ Protease is highly site-specific, highly active and significantly more stable than native TEV protease, resulting in enhanced long-term activity.

  • Highly specific cleavage activity
  • Enhanced enzyme stability for prolonged protease activity
  • Activity over a broad temperature (4°C to 30°C) and pH (6.0 to 8.5) range
  • Six-histidine sequence to facilitate its removal from the digested protein sample
  • Greater than 85% single-band purity with no nonspecific protease contamination

  • Application
    • Incubation with AcTEV™ Protease releases the protein of interest from the fusion tag
    • This is an effective way to remove solubility, secretion, detection, and purification tags from recombinant protein
    Enzyme specifications
    • Purified from E. coli expressing the AcTEV™ Protease gene
  • Unit definition
    • One unit of AcTEV™ Protease cleaves 85% of a 3μg control substrate in 1 hr at 30°C
  • Unit reaction conditions
    • 50mM Tris-HCl (pH 8.0), 0.5mM EDTA, 1mM DTT, 3μg control substrate, and 1 unit enzyme in 30μL for 1 hr at 30°C AcTEV™ Protease is functionally tested for the absence of any nonspecific protease activity

    GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1,000 Units
Content And Storage AcTEV™ Protease is supplied with a vial of 20X TEV buffer [1 M Tris-HCl (pH 8.0), 10 mM EDTA] and a vial of 100 mM DTT.

Store at -20° C. Guaranteed stable for 1 year when properly stored.
Shipping Condition Dry Ice
Enzyme TEV Protease
Compatible Buffer TEV Buffer
Optimal Reaction Temperature +4°C to +30°C
Quantity 1000 U
Product Type Protease
Product Line AcTEV

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.