Learn More
Invitrogen™ Novex™ 8%, Tris-Glycine, Protein Gel, 12 + 2-well, w/adapters
Separation Range: 40 to 200 kd; Separation Type: Denaturing, Native; For Use With: Bio-Rad Criterion, Midi Gel Tank, XCell4 SureLock Midi
Brand: Invitrogen™ WT0081A
Code : 50
Additional Details : Weight : 0.98000kg
Includes: One box of 10 gels, adapters
Description
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards; Usable for native and denaturing protein assays
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 45μL sample + 15μL standards; 1D Well
Compatibility: Standard multi-channel pipettor
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
XCell4 SureLock Midi-Cell, Bio-Rad Criterion Cell | |
Tris-Glycine | |
Protein | |
Denaturing, Native | |
1 Box | |
13 cm |
8% | |
Homogeneous Page | |
40 to 200 kDa | |
12 + 2-well | |
8 cm |
For Research Use Only. Not for use in diagnostic procedures.