missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Novex™ Tris-Glycine Plus Midi Protein Gels, 8 to 16% ,1.0 mm

Product Code. 15791819
Click to view available options
Wells:
12 + 2-well
12 + 2-well, w/adapters
20-well
20-well, w/adapters
26-well
26-well, w/adapters
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15791819

Brand: Novex™ WXP81626BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novex Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based on traditional Laemmli protein electrophoresis technology, allowing the use of Laemmli sample and running buffers.

Novex Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based on traditional Laemmli protein electrophoresis technology, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus Midi gels are a 1.0-mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples.

Features of Novex Tris-Glycine Plus gels include:
Improved shelf life—store gels for up to 12 months at 4°C
Flexibility—use for native and denaturing protein assays
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine Plus Midi gels come in fixed concentrations of 10% and 12%, as well as gradient concentrations of 4–12%, 4–20%, and 8–16%. Select from our many well formats, including 12+2-, 20-, and 26-well.

Run your proteins in native or denatured form
Novex Tris-Glycine Plus gels do not contain SDS and so can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26754). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672). The gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion Cell using our adapters (WA0999).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) when performing traditional wet transfer. Rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Recommended Storage

Store in refrigerator (2-8°C). Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Adapters No Adapters
For Use With (Equipment) XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage 8 to 16%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
Sample Loading Volume Up to 15 μL
Separation Range 6 to 250 kDa
Separation Type Denaturing, Native
Wells 26-well
Mode of Separation Molecular Weight
Product Line Novex
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Storage Requirements Store at 2°C to 8°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 13 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.