missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 1-well

Product Code. 12065675
Change view
Click to view available options
Quantity:
10 gels (1 box)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12065675 10 gels (1 box) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 12065675 Supplier Invitrogen™ Supplier No. EC6021BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One box of 10 gels

Separation Range: 20 to 200 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini

  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Individually packaged gels for optimal experiment design
  • Compatible with most protein standards; usable for native and denaturing protein assays
  • For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase
  • Run proteins in native or denatured form
  • Do not contain SDS and can be used to run proteins in native or in denatured form

Sample Loading Volume: 700μL; 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks.
Gel Percentage 4 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Sample Loading Volume 700 μL
Separation of Protein
Separation Range 20 to 200 kDa
Separation Type Denaturing, Native
Wells 1-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Thickness 1.0 mm
Well Design 1D Well
Length (Metric) 8 cm
Length (Metric) Gel 8 cm
Width (Metric) 8 cm
Width (Metric) Gel 8 cm
Product Line Novex
Product Type Gradient Page
For Use With (Equipment) XCell SureLock Mini-Cell
System Type Novex
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.