missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ 18%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 10 well

Separation Range: 6 to 40 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Systems

Brand:  Invitrogen™ EC6505BOX

Product Code. 12040106

  • £151.00 / Pack of 10
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: One box of 10 gels
This item is not returnable. View return policy

Description

Description

This product is being replaced by Catalog number XP00180BOX and will no longer be available after May 31, 2016.

  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Individually packaged gels for optimal experiment design
  • Compatible with most protein standards; usable for native and denaturing protein assays
  • For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase
  • Run proteins in native or denatured form
  • Do not contain SDS and can be used to run proteins in native or in denatured form

Sample Loading Volume: 25μL; 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

18%
Tris-Glycine
6 to 40 kd
10-well
8 cm
8 cm
Mini
Protein
Denaturing, Native
Each
8 cm
8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only. Not for use in diagnostic procedures.