missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ 12%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 15-well
Click to view available options
Wells:
15-well
Unit Size:
Pack of 10
Description
Includes
One box of 10 gels
This product is being replaced by Catalog number XP00125BOX and will no longer be available after May 31, 2016.
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards
- Usable for native and denaturing protein assays
- For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase
- Run proteins in native or denatured form
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 15 μL; 1D Well
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| For Use With (Equipment) | XCell SureLock Mini-Cell |
| Gel Percentage | 12% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Tris-Glycine |
| Product Type | Homogeneous Page |
| Separation of | Protein |
| Separation Range | 20 to 250 kDa |
| Separation Type | Denaturing, Native |
| Wells | 15-well |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction