missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nova2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85393-0.1ML
This item is not returnable.
View return policy
Description
Nova2 Polyclonal specifically detects Nova2 in Human samples. It is validated for Western Blot.
Specifications
| Nova2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4858 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| neuro-oncological ventral antigen 2ANOVANOVA3Astrocytic NOVA1-like RNA-binding protein, neuro-oncological ventral antigen 3, RNA-binding protein Nova-2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human Nova2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur