missing translation for 'onlineSavingsMsg'
Learn More

NOVA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55002-1-AP-20UL

 View more versions of this product

Product Code. 16896425

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody is specific to NOVA2.

Paraneoplastic opsoclonus myoclonus ataxia (POMA) is a neurologic disorder thought to be mediated by an autoimmune attack against onconeural disease antigens that are expressed by gynecologic or lung tumors and by neurons. Nova-1 and -2 are both POMA disease antigens. The autoimmune attack against Nova-2 in POMA patients results in latent cancer, reduced inhibitory control of movements, and dementia. Nova-2 is closely related to Nova-1, and is expressed at high levels in neurons during development and at lower levels in the adult lung. In the postnatal mouse brain, Nova-2 is expressed in a pattern that is largely reciprocal with Nova-1, including high levels of Nova-2 expression in the neocortex and hippocampus. Both Novas are RNA binding proteins, and Nova-2 binds RNA with high affinity and with sequence specificity that differs from Nova-1. They regulate neuronal pre-messenger RNA splicing by directly binding to RNA. Nova-2 target genes function either in the synapse or in axon guidance indicating that Nova-2 is a major regulator of synaptic function.
Specifications

Specifications

NOVA2
Polyclonal
Unconjugated
NOVA2
ANOVA, NOVA2, NOVA3, RNA binding protein Nova 2
Rabbit
Antigen Affinity Chromatography
RUO
292681, 384569, 4858
-20°C
Liquid
Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UNW9
Nova2
Peptide
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.