missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOVA1 Recombinant Rabbit Monoclonal Antibody (K01_1U87)

Product Code. 30830275
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830275 50 μL
30829392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830275 Supplier Invitrogen™ Supplier No. MA565183
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NOVA1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOVA1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1U87
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene NOVA1
Gene Accession No. P51513, Q80WA4, Q9JKN6
Gene Alias 9430099M15Rik; G630039L02; neuro-oncological ventral antigen 1; NOVA alternative splicing regulator 1; Nova1; Nova-1; Onconeural ventral antigen 1; paraneoplastic Ri antigen; RNA-binding protein Nova-1; ventral neuron-specific protein 1
Gene Symbols NOVA1
Host Species Rabbit
Immunogen A synthetic peptide of human Nova1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298992, 4857, 664883
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.