missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nova1 Antibody (S06-8K6), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | Nova1 |
|---|---|
| Clone | S06-8K6 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Sandwich ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA, Immunofluorescence, Sandwich ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667943
|
Novus Biologicals
NBP3-19720-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656463
|
Novus Biologicals
NBP3-19720-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nova1 Monoclonal antibody specifically detects Nova1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISASpecifications
| Nova1 | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunofluorescence, Sandwich ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| neuro-oncological ventral antigen 1Nova-1, Onconeural ventral antigen 1, Paraneoplastic Ri antigen, RNA-binding protein Nova-1, Ventral neuron-specific protein 1 | |
| A synthetic peptide of human Nova1 (Uniprot # P51513) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-8K6 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Sandwich ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 4857 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title