missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nova1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00221
This item is not returnable.
View return policy
Description
Nova1 Polyclonal specifically detects Nova1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Nova1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| neuro-oncological ventral antigen 1Nova-1, Onconeural ventral antigen 1, Paraneoplastic Ri antigen, RNA-binding protein Nova-1, Ventral neuron-specific protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4857 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.01 - 0.03 ug/mL, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:64000 | |
| NP_002506.2 | |
| NOVA1 | |
| Peptide with sequence C-REMPQNVAKTEPVS corresponding to internal region according to NP_002506.2 NP_006480.2 NP_006482.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize all three reported isoforms (NP_002506.2 ; NP_006480.2 ; NP_006482.1) | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction