missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nova-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571356
This item is not returnable.
View return policy
Description
Nova-2 Polyclonal Antibody for Western Blot
Paraneoplastic opsoclonus myoclonus ataxia (POMA) is a neurologic disorder thought to be mediated by an autoimmune attack against onconeural disease antigens that are expressed by gynecologic or lung tumors and by neurons. Nova-1 and -2 are both POMA disease antigens. The autoimmune attack against Nova-2 in POMA patients results in latent cancer, reduced inhibitory control of movements, and dementia. Nova-2 is closely related to Nova-1, and is expressed at high levels in neurons during development and at lower levels in the adult lung. In the postnatal mouse brain, Nova-2 is expressed in a pattern that is largely reciprocal with Nova-1, including high levels of Nova-2 expression in the neocortex and hippocampus. Both Novas are RNA binding proteins, and Nova-2 binds RNA with high affinity and with sequence specificity that differs from Nova-1. They regulate neuronal pre-messenger RNA splicing by directly binding to RNA. Nova-2 target genes function either in the synapse or in axon guidance indicating that Nova-2 is a major regulator of synaptic function.
Specifications
| Nova-2 | |
| Polyclonal | |
| Unconjugated | |
| NOVA2 | |
| ANOVA; astrocytic NOVA1-like RNA-binding protein; Gm1424; neuro-oncological ventral antigen 2; neuro-oncological ventral antigen 3; NOVA alternative splicing regulator 2; NOVA2; NOVA3; RNA-binding protein Nova-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4858 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UNW9 | |
| NOVA2 | |
| synthetic peptide directed towards the middle region of human Nova-2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction