missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOTUM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520788
This item is not returnable.
View return policy
Description
A suggested positive control is human kidney tissue lysate. PA5-20788 can be used with blocking peptide PEP-0902.
Genetic studies in Drosophila have revealed a novel secreted antagonist for Wingless (Wg) signaling, referred to as Notum/Wingful (Wf). Wf/Notum encodes a protein of 430 amino acid residues, which is related to plant pectin acetylesterases of the a/beta-hydrolase superfamily. The main target of Notum action appears to be cell surface proteoglycans Dally and/or Dally-like (Dlp). Notum governs the extracellular Wg morphogen distribution via Dally and Dlp. Also, like some vertebrate Wnt antagonists, notum expression is activated by Wg signaling, revealing the existence of a feedback control mechanism in which Wg modifies its own gradient distribution. In vertebrate, proteoglycans related to Dally and Dlp likely play important roles in Wnt signaling which encodes a glypican that is required for Wnt-11 signaling. A single ortholog of Notum exists in the mouse and the human genome, implying that a similar regulatory mechanism may exist in vertebrates.
Specifications
| NOTUM | |
| Polyclonal | |
| Unconjugated | |
| NOTUM | |
| [Wnt protein] O-palmitoleoyl-L-serine hydrolase; 5730593N15Rik; hNOTUM; NOTUM; notum pectinacetylesterase homolog; notum pectinacetylesterase homolog (Drosophila); NOTUM, palmitoleoyl-protein carboxylesterase; OK/SW-CL.30; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog; RGD1307119 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147111, 303743, 77583 | |
| 4°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6P988, Q8R116 | |
| NOTUM | |
| A 19 amino acid peptide near the amino terminus of human NOTUM. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction